Assay Design



Gene Transcripts

Gene Symbol
gatd1
Entrez Gene ID
100148360
Gene Name
glutamine amidotransferase like class 1 domain containing 1
Gene Aliases
pddc1, zgc:123196
Chromosome Location
Chr.25: 3801227 - 3808635 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037118.1 NP_001032195.1 6 - 7 729 115
GenBank mRNA BC085644.1 - 5 - 6 2109 115
BC107609.1 - 6 - 7 729 115

View Full Product Details