Assay Design



Gene Transcripts

Gene Symbol
arl3b
Entrez Gene ID
797451
Gene Name
ADP ribosylation factor like GTPase 3b
Gene Aliases
arl3, si:ch211-208d15.4, wu:fj22g05
Chromosome Location
Chr.12: 100732 - 107057 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044908.2 NP_001038373.1 4 - 5 356 127
GenBank mRNA BC124704.1 - 4 - 5 356 127
BC164965.1 - 3 - 4 350 127

View Full Product Details