Assay Design



Gene Transcripts

Gene Symbol
btr12
Entrez Gene ID
767681
Gene Name
bloodthirsty-related gene family, member 12
Gene Aliases
zgc:153732
Chromosome Location
Chr.7: 73668061 - 73685576 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076619.1 NP_001070087.1 2 - 3 730 88
XM_017356912.2 XP_017212401.1 2 - 3 846 88
GenBank mRNA BC124441.1 - 2 - 3 730 88

View Full Product Details