Assay Design



Gene Transcripts

Gene Symbol
socs5a
Entrez Gene ID
768142
Gene Name
suppressor of cytokine signaling 5a
Gene Aliases
socs5, zgc:153506
Chromosome Location
Chr.10: 25312318 - 25312647 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113758.2 NP_001107230.1 1 - 2 381 92
GenBank mRNA EF195763.1 - 1 - 2 381 92

View Full Product Details