Assay Design



Gene Transcripts

Gene Symbol
hs3st3l
Entrez Gene ID
100008592
Gene Name
heparan sulfate (glucosamine) 3-O-sulfotransferase 3-like
Gene Aliases
3-OST-6, 3OST6, HS3st6
Chromosome Location
Chr.12: 415804 - 431149 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081440.1 NP_001074909.1 1 - 2 378 99
GenBank mRNA BC163067.1 - 1 - 2 478 99
BC163072.1 - 1 - 2 478 99
DQ812991.1 - 1 - 2 378 99

View Full Product Details