Assay Design



Gene Transcripts

Gene Symbol
mfsd6a
Entrez Gene ID
100034426
Gene Name
major facilitator superfamily domain containing 6a
Gene Aliases
mfsd6, si:dkey-188p4.2
Chromosome Location
Chr.22: 12911236 - 12931202 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083056.1 NP_001076525.1 3 - 4 1660 64
XM_005161679.4 XP_005161736.1 4 - 5 2629 64
XM_021469281.1 XP_021324956.1 3 - 4 1660 64

View Full Product Details