Assay Design



Gene Transcripts

Gene Symbol
rhcga
Entrez Gene ID
100038797
Gene Name
Rh family, C glycoprotein a
Gene Aliases
rhcg2b, zgc:162132
Chromosome Location
Chr.7: 14291349 - 14302716 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089577.1 NP_001083046.1 10 - 11 1462 73
GenBank mRNA AB286867.1 - 10 - 11 1487 73
BC139524.1 - 10 - 11 1462 73

View Full Product Details