Assay Design



Gene Transcripts

Gene Symbol
shda
Entrez Gene ID
561247
Gene Name
Src homology 2 domain containing transforming protein D, a
Gene Aliases
si:ch211-137c4.3
Chromosome Location
Chr.2: 52065884 - 52093505 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110835.1 NP_001104305.1 3 - 4 513 81

View Full Product Details