Assay Design



Gene Transcripts

Gene Symbol
pdp1
Entrez Gene ID
558728
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 1
Gene Aliases
im:7157162, zgc:174908
Chromosome Location
Chr.16: 26816756 - 26820634 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111158.1 NP_001104628.1 1 - 2 96 91
GenBank mRNA BC155791.1 - 1 - 2 96 91

View Full Product Details