Assay Design



Gene Transcripts

Gene Symbol
march8
Entrez Gene ID
567818
Gene Name
membrane-associated ring finger (C3HC4) 8
Gene Aliases
zgc:171495
Chromosome Location
Chr.13: 17752591 - 18011412 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009307237.3 XP_009305512.1 2 - 3 478 119
XM_009307239.3 XP_009305514.1 2 - 3 455 119
GenBank mRNA BC153940.1 - 2 - 3 267 119

View Full Product Details