Assay Design
Gene Transcripts
- Gene Symbol
- myh7
- Entrez Gene ID
-
30616
- Gene Name
- myosin heavy chain 7
- Gene Aliases
-
mg:cb02g011, smyhc3, vmhc
- Chromosome Location
-
Chr.2: 24256521 - 24269913 on Build GRCz11
- Gene Symbol
- myh7bb
- Entrez Gene ID
-
560945
- Gene Name
- myosin, heavy chain 7B, cardiac muscle, beta b
- Gene Aliases
-
myh7, si:ch211-209j10.4
- Chromosome Location
-
Chr.23: 18722680 - 18757172 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EB891720.1 |
- |
1 - 2 |
66 |
65 |
View Full Product Details