Assay Design



Gene Transcripts

Gene Symbol
st8sia3
Entrez Gene ID
553510
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
Gene Aliases
siat 8C
Chromosome Location
Chr.21: 1780790 - 1799265 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113432.2 NP_001106903.1 3 - 4 1081 69
GenBank mRNA BC163895.1 - 3 - 4 921 69
BC163914.1 - 3 - 4 921 69
DQ853412.1 - 3 - 4 1083 69

View Full Product Details