Assay Design



Gene Transcripts

Gene Symbol
mfsd4ab
Entrez Gene ID
558146
Gene Name
major facilitator superfamily domain containing 4Ab
Gene Aliases
im:7150270, mfsd4b2, zgc:175151
Chromosome Location
Chr.23: 18103103 - 18114890 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114416.1 NP_001107888.1 7 - 8 1316 80
GenBank mRNA BC155330.1 - 7 - 8 1316 80

View Full Product Details