Assay Design



Gene Transcripts

Gene Symbol
slc26a6l
Entrez Gene ID
559448
Gene Name
solute carrier family 26 member 6, like
Gene Aliases
SLC26A6, zgc:175226
Chromosome Location
Chr.6: 40723554 - 40732692 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114417.1 NP_001107889.1 14 - 15 1747 69
GenBank mRNA BC155340.1 - 14 - 15 1747 69
FJ170818.1 - 14 - 15 1746 69

View Full Product Details