Assay Design



Gene Transcripts

Gene Symbol
ahr1a
Entrez Gene ID
246224
Gene Name
aryl hydrocarbon receptor 1a
Gene Aliases
AHR2, ahr1, ahra, zfAHR1
Chromosome Location
Chr.16: 20009938 - 20038280 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131028.2 NP_571103.2 8 - 9 1333 91
XM_021466612.1 XP_021322287.1 8 - 9 1350 91
GenBank mRNA AF258854.1 - 8 - 9 1333 91

View Full Product Details