Assay Design



Gene Transcripts

Gene Symbol
bfb
Entrez Gene ID
30489
Gene Name
complement component bfb
Gene Aliases
Bf-2
Chromosome Location
Chr.21: 27437934 - 27443999 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131241.1 NP_571316.1 6 - 7 859 114
XM_021468750.1 XP_021324425.1 6 - 7 906 114
GenBank mRNA AF047412.1 - 4 - 5 557 114
BC076051.1 - 6 - 7 859 114

View Full Product Details