Assay Design
Gene Transcripts
- Gene Symbol
- rag2
- Entrez Gene ID
-
30658
- Gene Name
- recombination activating gene 2
- Gene Aliases
-
rag-2, zgc:136743
- Chromosome Location
-
Chr.25: 9010425 - 9013963 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_131385.3 |
NP_571460.2 |
1 - 2 |
146 |
74 |
GenBank mRNA |
BC115249.1 |
- |
1 - 2 |
73 |
74 |
View Full Product Details