Assay Design



Gene Transcripts

Gene Symbol
nitr4a
Entrez Gene ID
60652
Gene Name
novel immune-type receptor 4a
Gene Aliases
dZ105L16.2, dZ105L16.3, dZ105L16.8, nitr, nitr4a_9
Chromosome Location
Chr.7: 17578061 - 17582292 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131656.1 NP_571731.1 2 - 3 399 124
NM_198344.3 NP_938158.3 2 - 3 405 124
GenBank mRNA AF318404.1 - 2 - 3 399 124

View Full Product Details