Assay Design



Gene Transcripts

Gene Symbol
mitfb
Entrez Gene ID
114833
Gene Name
melanocyte inducing transcription factor b
Gene Aliases
-
Chromosome Location
Chr.23: 709269 - 764135 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131847.2 NP_571922.2 4 - 5 731 111
XM_021469718.1 XP_021325393.1 3 - 4 757 111
GenBank mRNA JN105118.1 - 4 - 5 731 111

View Full Product Details