Assay Design



Gene Transcripts

Gene Symbol
dffb
Entrez Gene ID
373130
Gene Name
DNA fragmentation factor, beta polypeptide (caspase-activated DNase)
Gene Aliases
CAD
Chromosome Location
Chr.8: 22277215 - 22282962 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194404.1 NP_919385.1 4 - 5 534 107
GenBank mRNA AF286179.1 - 4 - 5 534 107
BC067602.1 - 4 - 5 546 107
BC092678.1 - 4 - 5 530 107
BC164850.1 - 4 - 5 544 107

View Full Product Details