Assay Design



Gene Transcripts

Gene Symbol
taf12
Entrez Gene ID
386637
Gene Name
TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
fl03a12, wu:fl03a12
Chromosome Location
Chr.16: 75497 - 81655 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198368.1 NP_938182.1 3 - 4 407 74
XM_009292430.3 XP_009290705.1 3 - 4 400 74
GenBank mRNA BC050160.1 - 3 - 4 433 74
BC056696.1 - 3 - 4 407 74
BC065878.1 - 3 - 4 387 74
BC152198.1 - 3 - 4 437 74

View Full Product Details