Assay Design



Gene Transcripts

Gene Symbol
depdc1a
Entrez Gene ID
100003959
Gene Name
DEP domain containing 1a
Gene Aliases
cb41, depdc1, sb:cb41, wu:fc05g03, zgc:55374
Chromosome Location
Chr.8: 65829 - 78118 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198872.1 NP_942573.1 4 - 5 790 101
XM_005166773.3 XP_005166830.1 4 - 5 791 101
GenBank mRNA BC044382.1 - 4 - 5 790 101
BC171596.1 - 4 - 5 742 101
BC171624.1 - 4 - 5 742 101

View Full Product Details