Assay Design



Gene Transcripts

Gene Symbol
psmd11b
Entrez Gene ID
322265
Gene Name
proteasome 26S subunit, non-ATPase 11b
Gene Aliases
psmd11, wu:fb55b06, zgc:56580, zgc:77763
Chromosome Location
Chr.12: 28349844 - 28363092 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199592.1 NP_955886.1 8 - 9 951 70
GenBank mRNA BC051618.1 - 8 - 9 951 70
BC063978.1 - 8 - 9 934 70

View Full Product Details