Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199645.1 | NP_955939.1 | 3 - 4 | 550 | 128 |
GenBank mRNA | BC057433.1 | - | 3 - 4 | 550 | 128 |
BC154323.1 | - | 3 - 4 | 566 | 128 | |
BC155749.1 | - | 3 - 4 | 532 | 128 |