Assay Design



Gene Transcripts

Gene Symbol
pigp
Entrez Gene ID
325316
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class P
Gene Aliases
dscr5, wu:fc66d08, zgc:73366
Chromosome Location
Chr.10: 110426 - 116036 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199712.2 NP_956006.2 4 - 5 344 118
XM_005172570.4 XP_005172627.1 4 - 5 797 118
GenBank mRNA BC059676.1 - 3 - 4 309 118

View Full Product Details