Assay Design



Gene Transcripts

Gene Symbol
ela2l
Entrez Gene ID
334304
Gene Name
elastase 2 like
Gene Aliases
ctrc, elastaseB, fi40a05, wu:fi40a05, zgc:111871, zgc:63744
Chromosome Location
Chr.8: 119959 - 123353 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199886.1 NP_956180.1 4 - 5 360 92
GenBank mRNA BC054136.1 - 4 - 5 360 92
BC093119.1 - 4 - 5 360 92
BC097021.1 - 4 - 5 359 92
BC163342.1 - 4 - 5 353 92

View Full Product Details