Assay Design
Gene Transcripts
- Gene Symbol
- ankmy2a
- Entrez Gene ID
-
334389
- Gene Name
- ankyrin repeat and MYND domain containing 2a
- Gene Aliases
-
fi46e08, wu:fi46e08, zgc:55491
- Chromosome Location
-
Chr.15: 34573311 - 34590820 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199894.1 |
NP_956188.1 |
9 - 10 |
1101 |
73 |
GenBank mRNA |
BC044410.1 |
- |
9 - 10 |
1101 |
73 |
View Full Product Details