Assay Design



Gene Transcripts

Gene Symbol
odf3b
Entrez Gene ID
335524
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
fj19d04, odf3l, si:dkeyp-1h4.4, trim35-30, wu:fj19d04, zgc:63985
Chromosome Location
Chr.18: 251070 - 257520 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199958.1 NP_956252.1 4 - 5 678 118
XM_021467596.1 XP_021323271.1 4 - 5 732 118
GenBank mRNA BC054587.1 - 4 - 5 678 118
BC163161.1 - 4 - 5 503 118
BC163187.1 - 4 - 5 503 118
BC164454.1 - 3 - 4 450 118

View Full Product Details