Assay Design
Gene Transcripts
- Gene Symbol
- tigarb
- Entrez Gene ID
-
393160
- Gene Name
- TP53 induced glycolysis regulatory phosphatase b
- Gene Aliases
-
c12orf5b, si:ch211-240j22.3, zgc:56074
- Chromosome Location
-
Chr.4: 5156117 - 5161877 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200191.1 |
NP_956485.1 |
4 - 5 |
393 |
70 |
GenBank mRNA |
BC045897.1 |
- |
4 - 5 |
393 |
70 |
View Full Product Details