Assay Design



Gene Transcripts

Gene Symbol
wdr45b
Entrez Gene ID
393210
Gene Name
WD repeat domain 45B
Gene Aliases
WIPI-3, wdr45l, zgc:56002
Chromosome Location
Chr.12: 40351 - 46985 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200240.2 NP_956534.2 8 - 9 1253 66
GenBank mRNA BC047802.1 - 8 - 9 909 66
BC164151.1 - 8 - 9 842 66

View Full Product Details