Assay Design
Gene Transcripts
- Gene Symbol
- cisd1
- Entrez Gene ID
-
393577
- Gene Name
- CDGSH iron sulfur domain 1
- Gene Aliases
-
si:dkey-18n13.4, zgc:63561
- Chromosome Location
-
Chr.13: 79006 - 85706 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200605.1 |
NP_956899.1 |
1 - 2 |
132 |
64 |
GenBank mRNA |
BC056783.1 |
- |
1 - 2 |
132 |
64 |
View Full Product Details