Assay Design



Gene Transcripts

Gene Symbol
sap18
Entrez Gene ID
393693
Gene Name
Sin3A-associated protein
Gene Aliases
zgc:73118
Chromosome Location
Chr.9: 136443 - 138358 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200720.2 NP_957014.1 1 - 2 189 100
GenBank mRNA BC059482.1 - 1 - 2 100
BC155197.1 - 1 - 2 137 100
EH440312.1 - 1 - 2 124 100

View Full Product Details