Assay Design
Gene Transcripts
- Gene Symbol
- arl5c
- Entrez Gene ID
-
393819
- Gene Name
- ADP-ribosylation factor-like 5C
- Gene Aliases
-
zgc:77751
- Chromosome Location
-
Chr.3: 16134376 - 16141895 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200846.1 |
NP_957140.1 |
4 - 5 |
554 |
60 |
GenBank mRNA |
BC062281.1 |
- |
4 - 5 |
554 |
60 |
View Full Product Details