Assay Design



Gene Transcripts

Gene Symbol
sobpb
Entrez Gene ID
100006088
Gene Name
sine oculis binding protein homolog (Drosophila) b
Gene Aliases
jxc1-B, si:ch211-152c2.4
Chromosome Location
Chr.4: 817095 - 861309 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114983.2 NP_001108455.1 4 - 5 565 57
XM_009299931.3 XP_009298206.1 4 - 5 872 57
XM_017354921.2 XP_017210410.1 5 - 6 2855 57
GenBank mRNA DQ503413.1 - 4 - 5 565 57

View Full Product Details