Assay Design



Gene Transcripts

Gene Symbol
edar
Entrez Gene ID
796370
Gene Name
ectodysplasin A receptor
Gene Aliases
fls
Chromosome Location
Chr.9: 56103173 - 56172757 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115064.2 NP_001108536.2 3 - 4 301 71
GenBank mRNA EF137867.1 - 3 - 4 301 71

View Full Product Details