Assay Design



Gene Transcripts

Gene Symbol
edar
Entrez Gene ID
796370
Gene Name
ectodysplasin A receptor
Gene Aliases
fls
Chromosome Location
Chr.9: 56103173 - 56172757 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115064.2 NP_001108536.2 6 - 7 656 73
GenBank mRNA EF137867.1 - 6 - 7 656 73

View Full Product Details