Assay Design



Gene Transcripts

Gene Symbol
pcdh18b
Entrez Gene ID
562031
Gene Name
protocadherin 18b
Gene Aliases
fj19f12, pcdh18, wu:fj19f12
Chromosome Location
Chr.14: 174892 - 182224 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115099.1 NP_001108571.1 2 - 3 2634 82
XM_005157329.4 XP_005157386.1 2 - 3 2969 82
GenBank mRNA AB297803.1 - 2 - 3 2634 82

View Full Product Details