Assay Design



Gene Transcripts

Gene Symbol
ptbp1b
Entrez Gene ID
321220
Gene Name
polypyrimidine tract binding protein 1b
Gene Aliases
cb363, id:ibd2901, im:7145323, sb:cb363, si:ch211-262i1.1, wu:fa04b05, wu:fb16f08, wu:fc58e03
Chromosome Location
Chr.11: 231474 - 253011 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122654.1 NP_001116126.1 8 - 9 838 81
XM_005155938.4 XP_005155995.1 7 - 8 767 81
XM_009305880.2 XP_009304155.1 8 - 9 787 81

View Full Product Details