Assay Design



Gene Transcripts

Gene Symbol
mxg
Entrez Gene ID
360148
Gene Name
myxovirus (influenza virus) resistance G
Gene Aliases
-
Chromosome Location
Chr.25: 19695429 - 19715137 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122971.1 NP_001116443.1 7 - 8 1010 92
XM_009297890.2 XP_009296165.1 7 - 8 1233 92
XM_021470456.1 XP_021326131.1 5 - 6 737 92
GenBank mRNA AJ544829.2 - 7 - 8 1010 92

View Full Product Details