Assay Design



Gene Transcripts

Gene Symbol
amd1
Entrez Gene ID
792097
Gene Name
adenosylmethionine decarboxylase 1
Gene Aliases
fb26e03, si:ch211-257g8.2, wu:fb26e03, zgc:55614
Chromosome Location
Chr.20: 50283 - 79278 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161601.1 NP_001155073.1 4 - 5 667 64
GenBank mRNA BC045404.1 - 4 - 5 646 64
BC163959.1 - 4 - 5 464 64

View Full Product Details