Assay Design
Gene Transcripts
- Gene Symbol
- bicc2
- Entrez Gene ID
-
402785
- Gene Name
- bicaudal C homolog 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 2941796 - 2979878 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203420.1 |
NP_981965.1 |
2 - 3 |
289 |
60 |
GenBank mRNA |
AY513238.1 |
- |
2 - 3 |
289 |
60 |
View Full Product Details