Assay Design
Gene Transcripts
- Gene Symbol
- rabl3
- Entrez Gene ID
-
403048
- Gene Name
- RAB, member of RAS oncogene family-like 3
- Gene Aliases
-
fb40b02, wu:fb40b02, zgc:100876
- Chromosome Location
-
Chr.9: 32649118 - 32652416 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205733.1 |
NP_991296.1 |
3 - 4 |
262 |
110 |
GenBank mRNA |
AY391440.1 |
- |
3 - 4 |
262 |
110 |
View Full Product Details