Assay Design



Gene Transcripts

Gene Symbol
tlr4bb
Entrez Gene ID
403132
Gene Name
toll-like receptor 4b, duplicate b
Gene Aliases
tlr4b
Chromosome Location
Chr.13: 18523654 - 18528859 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212813.2 NP_997978.2 2 - 3 443 147
GenBank mRNA AY388400.1 - 2 - 3 247 147
BC068358.1 - 2 - 3 436 147

View Full Product Details