Assay Design



Gene Transcripts

Gene Symbol
tmbim4
Entrez Gene ID
406412
Gene Name
transmembrane BAX inhibitor motif containing 4
Gene Aliases
wu:fb34a09, zgc:64112
Chromosome Location
Chr.4: 12971250 - 12978931 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213138.2 NP_998303.2 3 - 4 445 96
GenBank mRNA BC057432.1 - 3 - 4 385 96
EH442390.1 - 3 - 4 452 96
EH490666.1 - 3 - 4 447 96

View Full Product Details