Assay Design



Gene Transcripts

Gene Symbol
nudt9
Entrez Gene ID
406661
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 9
Gene Aliases
fu33g07, sb:cb392, wu:fj16b09, wu:fu33g07, zgc:63924
Chromosome Location
Chr.3: 26223374 - 26232773 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213352.2 NP_998517.2 6 - 7 829 114
GenBank mRNA BC057417.1 - 6 - 7 114
BC152216.1 - 6 - 7 770 114
EH523281.1 - 2 - 3 637 114

View Full Product Details