Assay Design
Gene Transcripts
- Gene Symbol
- exo1
- Entrez Gene ID
-
406778
- Gene Name
- exonuclease 1
- Gene Aliases
-
fc16e04, wu:fc16e04, zgc:55521
- Chromosome Location
-
Chr.17: 282623 - 290235 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213469.1 |
NP_998634.1 |
2 - 3 |
255 |
101 |
GenBank mRNA |
BC044187.1 |
- |
2 - 3 |
255 |
101 |
View Full Product Details