Assay Design
Gene Transcripts
- Gene Symbol
- tax1bp3
- Entrez Gene ID
-
406779
- Gene Name
- Tax1 (human T-cell leukemia virus type I) binding protein 3
- Gene Aliases
-
wu:fb37e03, wu:fc23g08, zgc:55513
- Chromosome Location
-
Chr.11: 6136260 - 6145158 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213470.1 |
NP_998635.1 |
3 - 4 |
348 |
74 |
GenBank mRNA |
BC042322.1 |
- |
3 - 4 |
348 |
74 |
View Full Product Details