Assay Design



Gene Transcripts

Gene Symbol
vcanb
Entrez Gene ID
323465
Gene Name
versican b
Gene Aliases
cspg2b, dcan, fb99b07, wu:fb99b07
Chromosome Location
Chr.10: 43188678 - 43262960 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214688.1 NP_999853.1 4 - 5 662 81
GenBank mRNA AB119257.1 - 4 - 5 662 81
BC162605.1 - 4 - 5 648 81

View Full Product Details