Assay Design



Gene Transcripts

Gene Symbol
myl12.1
Entrez Gene ID
326719
Gene Name
myosin, light chain 12, genome duplicate 1
Gene Aliases
bing3y, hm:zeh0157, mlc-4, mlc12.1, myl9l, wu:fa93e09, wu:fe06h06, zeh0157, zgc:65791
Chromosome Location
Chr.7: 71764701 - 71772711 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214699.2 NP_999864.1 3 - 4 629 91
GenBank mRNA BC056526.1 - 3 - 4 408 91
EH434629.1 - 3 - 4 456 91
EH557418.1 - 1 - 2 813 91

View Full Product Details