Assay Design



Gene Transcripts

Gene Symbol
tmem47
Entrez Gene ID
406640
Gene Name
transmembrane protein 47
Gene Aliases
wu:fb99e01, zgc:63990
Chromosome Location
Chr.8: 18565904 - 18582998 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214763.2 NP_999928.1 2 - 3 587 68
GenBank mRNA BC057424.1 - 2 - 3 511 68
BC164961.1 - 2 - 3 398 68
EH543648.1 - 2 - 3 624 68

View Full Product Details